Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PILRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013439.2 | NP_038467.2 | 7 | 1143 | ||
NM_178272.1 | NP_840056.1 | 6 | 924 | |||
NM_178273.1 | NP_840057.1 | 5 | 890 | |||
GenBank mRNA | AJ400841.1 | - | 7 | 1084 | ||
AJ400842.1 | - | 6 | 841 | |||
AJ400843.1 | - | 5 | 806 | |||
BC017812.1 | - | 6 | 873 | 4690881 |