Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAZ2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289975.1 | NP_001276904.1 | 36 | 7617 | ||
NM_013450.3 | NP_038478.2 | 37 | 7725 | |||
XM_011511044.1 | XP_011509346.1 | 35 | 7352 | |||
XM_011511051.1 | XP_011509353.1 | 35 | 7241 | |||
XM_017003923.1 | XP_016859412.1 | 36 | 7454 | |||
XM_017003927.1 | XP_016859416.1 | 37 | 7606 | |||
XM_017003929.1 | XP_016859418.1 | 37 | 7493 | |||
XM_017003932.1 | XP_016859421.1 | 34 | 7157 | |||
XM_017003936.1 | XP_016859425.1 | 35 | 7170 | |||
GenBank mRNA | AB032255.1 | - | 32 | 7098 | ||
AB040909.2 | - | 36 | 7585 | |||
AK000386.1 | - | 1 | 1724 | |||
AK023566.1 | - | 1 | 1872 | |||
AL080173.1 | - | 8 | 2165 |