Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013451.3 | NP_038479.1 | 16 | 1469 | ||
NM_133337.2 | NP_579899.1 | 16 | 1469 | |||
XM_005269693.4 | XP_005269750.1 | 16 | 1648 | |||
XM_005269694.4 | XP_005269751.1 | 16 | 1648 | |||
XM_011539632.2 | XP_011537934.1 | 17 | 1842 | |||
XM_017016068.1 | XP_016871557.1 | 15 | 1229 | |||
XM_017016069.1 | XP_016871558.1 | 15 | 1274 | |||
XM_017016070.1 | XP_016871559.1 | 16 | 1656 | |||
GenBank mRNA | AB033033.2 | - | 16 | 1346 | ||
AF182316.1 | - | 16 | 1442 | |||
AF207990.1 | - | 16 | 1346 | |||
BC052617.1 | - | 16 | 1434 | 6422729 |