Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013451.3 | NP_038479.1 | 29 | 3210 | ||
NM_133337.2 | NP_579899.1 | 28 | 3171 | |||
XM_005269693.4 | XP_005269750.1 | 29 | 3389 | |||
XM_005269694.4 | XP_005269751.1 | 28 | 3350 | |||
XM_011539632.2 | XP_011537934.1 | 30 | 3583 | |||
XM_017016068.1 | XP_016871557.1 | 28 | 2970 | |||
XM_017016069.1 | XP_016871558.1 | 28 | 3015 | |||
XM_017016070.1 | XP_016871559.1 | 29 | 3397 | |||
GenBank mRNA | AB033033.2 | - | 29 | 3087 | ||
AF182316.1 | - | 29 | 3183 | |||
AF207990.1 | - | 28 | 3048 | |||
AL096713.1 | - | 9 | 1227 | |||
BC052617.1 | - | 29 | 3175 | 6422729 |