Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014030.3 | NP_054749.2 | 7, 8 | 983 | ||
XM_011524685.1 | XP_011522987.1 | 7, 8 | 824 | |||
GenBank mRNA | AF124490.1 | - | 7, 8 | 783 | ||
AK024826.1 | - | 2, 3 | 138 | |||
BC067358.1 | - | 7, 8 | 983 | |||
BQ424386.1 | - | 6, 7 | 622 | |||
BX537553.1 | - | 1 | 559 | |||
BX537554.1 | - | 1 | 559 | |||
BX648097.1 | - | 1 | 562 |