Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085454.1 | NP_001078923.1 | 10 | 1064 | ||
NM_014030.3 | NP_054749.2 | 9 | 1037 | |||
XM_011524684.1 | XP_011522986.1 | 10 | 905 | |||
XM_011524685.1 | XP_011522987.1 | 9 | 878 | |||
GenBank mRNA | AB209537.1 | - | 10 | 863 | ||
AF124490.1 | - | 9 | 837 | |||
AK024826.1 | - | 4 | 192 | |||
AK294785.1 | - | 11 | 942 | |||
AK297443.1 | - | 7 | 506 | |||
AK299932.1 | - | 10 | 908 | |||
BC067358.1 | - | 9 | 1037 | |||
BQ424386.1 | - | 8 | 676 | |||
BX537553.1 | - | 2 | 613 | |||
BX537554.1 | - | 2 | 613 | |||
BX648097.1 | - | 2 | 616 |