Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085454.1 | NP_001078923.1 | 21 | 3720 | ||
NM_014030.3 | NP_054749.2 | 20 | 3693 | |||
XM_011524684.1 | XP_011522986.1 | 21 | 3582 | |||
XM_011524685.1 | XP_011522987.1 | 20 | 3555 | |||
GenBank mRNA | AF124490.1 | - | 20 | 3499 | ||
AK000863.1 | - | 4 | 1720 | |||
AK024826.1 | - | 15 | 2845 | |||
BC005031.1 | - | 1 | 1155 | |||
BC006227.2 | - | 4 | 1727 | |||
BC018998.2 | - | 3 | 1635 | |||
BC032098.1 | - | 3 | 1644 | |||
BC048196.1 | - | 5 | 1795 | |||
BX537553.1 | - | 12 | 3429 | |||
BX537554.1 | - | 12 | 3429 | |||
BX648097.1 | - | 12 | 3431 |