Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014049.4 | NP_054768.2 | 11 | 1266 | ||
NR_033426.1 | 11 | 1442 | ||||
XM_017006264.1 | XP_016861753.1 | 11 | 1430 | |||
XM_017006265.1 | XP_016861754.1 | 5 | 416 | |||
GenBank mRNA | AB209427.1 | - | 11 | 2963 | ||
AF327351.1 | - | 11 | 1096 | |||
AK022568.1 | - | 10 | 1192 | |||
AK024012.1 | - | 11 | 1110 | |||
AK027186.1 | - | 1 | 88 | |||
AK130311.1 | - | 11 | 1137 | |||
AK294597.1 | - | 11 | 934 | |||
AK297952.1 | - | 11 | 1310 | |||
AK310644.1 | - | 10 | 1039 | |||
BC001817.2 | - | 4 | 230 | |||
BC007970.2 | - | 11 | 1143 | 4304209 | ||
BC013354.2 | - | 11 | 1120 | 3935925 |