Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014049.4 | NP_054768.2 | 17, 18 | 1965 | ||
NR_033426.1 | 17, 18 | 2141 | ||||
GenBank mRNA | AB209427.1 | - | 15, 16 | 4969 | ||
AF078854.1 | - | 5, 6 | 497 | |||
AF327351.1 | - | 17, 18 | 1795 | |||
AK022568.1 | - | 16, 17 | 1891 | |||
AK024012.1 | - | 17, 18 | 1809 | |||
AK027186.1 | - | 6, 7 | 1387 | |||
AK130311.1 | - | 17, 18 | 1836 | |||
AK297952.1 | - | 17, 18 | 2009 | |||
BC001817.2 | - | 10, 11 | 929 | |||
BC007970.2 | - | 17, 18 | 1842 | 4304209 | ||
BC013354.2 | - | 17, 18 | 1819 | 3935925 |