Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM230 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009923.1 | NP_001009923.1 | 5 | 1444 | ||
NM_001009924.1 | NP_001009924.1 | 5 | 1662 | |||
NM_001009925.1 | NP_001009925.1 | 4 | 1338 | |||
NM_014145.4 | NP_054864.3 | 4 | 1569 | |||
XM_005260713.3 | XP_005260770.1 | 4 | 1536 | |||
XM_011529227.2 | XP_011527529.1 | 4 | 1559 | |||
XM_011529229.2 | XP_011527531.1 | 5 | 1584 | |||
XM_017027836.1 | XP_016883325.1 | 4 | 1321 | |||
GenBank mRNA | AK055700.1 | - | 5 | 1618 | ||
AL357536.1 | - | 2 | 1032 | |||
AY359115.1 | - | 4 | 1461 | |||
BC009768.1 | - | 4 | 1322 | 4092724 | ||
BC009769.1 | - | 4 | 1328 | 4132400 | ||
BC009770.1 | - | 4 | 1322 | 4249231 | ||
BC011990.1 | - | 4 | 1310 | 3857616 | ||
BC015113.1 | - | 4 | 1273 | 4049350 | ||
BC070212.1 | - | 5 | 1300 | 4687557 | ||
BC110408.1 | - | 4 | 1300 | 5740520 |