Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014161.4 | NP_054880.2 | 3, 4 | 610 | ||
NR_134860.1 | 4, 5 | 730 | ||||
NR_134861.1 | 3, 4 | 687 | ||||
NR_134862.1 | 3, 4 | 610 | ||||
NR_134863.1 | 3, 4 | 594 | ||||
NR_134864.1 | 3, 4 | 493 | ||||
GenBank mRNA | AF161556.1 | - | 3, 4 | 588 | ||
AK311391.1 | - | 4, 5 | 607 | |||
AL136633.1 | - | 3, 4 | 560 | |||
BC001623.1 | - | 3, 4 | 581 | 3357333 | ||
BF797811.1 | - | 3, 4 | 494 | |||
BM924328.1 | - | 3, 4 | 703 | |||
BU601065.1 | - | 3, 4 | 551 | |||
BU932963.1 | - | 3, 4 | 599 | |||
CD675657.1 | - | 3, 4 | 493 | |||
CR533491.1 | - | 3, 4 | 468 |