Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014175.3 | NP_054894.1 | 3 | 472 | ||
XM_017013362.1 | XP_016868851.1 | 3 | 483 | |||
GenBank mRNA | AF161494.1 | - | 3 | 464 | ||
AK312832.1 | - | 3 | 436 | |||
BC000891.1 | - | 3 | 472 | 3463430 |