Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001326380.1 | NP_001313309.1 | 4 | 932 | ||
NM_001326381.1 | NP_001313310.1 | 4 | 1045 | |||
NM_014320.2 | NP_055135.1 | 4 | 1163 | |||
XM_017010631.1 | XP_016866120.1 | 4 | 1018 | |||
GenBank mRNA | AK057928.1 | - | 4 | 1078 |