Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014321.3 | NP_055136.1 | 3, 4 | 427 | ||
NR_037620.1 | 3, 4 | 486 | ||||
XM_011522978.2 | XP_011521280.1 | 3, 4 | 417 | |||
GenBank mRNA | AF139658.1 | - | 3, 4 | 415 | ||
AK021946.1 | - | 3, 4 | 388 | |||
AK024019.1 | - | 3, 4 | 421 | |||
AK024077.1 | - | 3, 4 | 414 | |||
AK310633.1 | - | 3, 4 | 450 | |||
BC039032.1 | - | 3, 4 | 407 | 5577396 | ||
BC063565.1 | - | 3, 4 | 396 | 3460593 | ||
BG822640.1 | - | 3, 4 | 398 |