Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CORO1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105237.2 | NP_001098707.1 | 11 | 1848 | ||
NM_001276471.1 | NP_001263400.1 | 11 | 1866 | |||
NM_014325.3 | NP_055140.1 | 11 | 1767 | |||
XM_011538125.1 | XP_011536427.1 | 7 | 1091 | |||
XM_017019121.1 | XP_016874610.1 | 12 | 2596 | |||
XM_017019122.1 | XP_016874611.1 | 11 | 2173 | |||
XM_017019123.1 | XP_016874612.1 | 12 | 2226 | |||
GenBank mRNA | AB030656.1 | - | 11 | 1736 | ||
AB209909.1 | - | 11 | 1722 | |||
AK096363.1 | - | 11 | 1866 | |||
AK223073.1 | - | 11 | 1735 | |||
AK297463.1 | - | 11 | 1600 | |||
AK311315.1 | - | 5 | 692 | |||
AK316065.1 | - | 11 | 1563 | |||
AL162070.1 | - | 11 | 1705 | |||
BC002342.2 | - | 11 | 1721 | 2822459 |