Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014335.2 | NP_055150.1 | 1 | 864 | ||
GenBank mRNA | AF092135.1 | - | 1 | 842 | ||
AF109873.1 | - | 1 | 842 | |||
AF349444.1 | - | 1 | 834 | |||
AK098383.1 | - | 2 | 753 | |||
AL109701.1 | - | 1 | 804 | |||
AL357456.1 | - | 1 | 792 | |||
BC114944.1 | - | 1 | 856 | 40084263 | ||
BC114946.1 | - | 1 | 856 | 40084266 |