Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC4E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014358.3 | NP_055173.1 | 6 | 680 | ||
XM_011520614.2 | XP_011518916.1 | 5 | 953 | |||
XM_011520615.1 | XP_011518917.1 | 5 | 484 | |||
GenBank mRNA | AB024718.1 | - | 6 | 666 | ||
AK312671.1 | - | 6 | 681 | |||
AY358499.1 | - | 6 | 620 | |||
BC000715.2 | - | 6 | 598 | 3507103 | ||
DA669951.1 | - | 2 | 132 |