Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIBCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014362.3 | NP_055177.2 | 8 | 859 | ||
NM_198047.2 | NP_932164.1 | 8 | 859 | |||
XM_011510953.1 | XP_011509255.1 | 8 | 808 | |||
XM_011510954.1 | XP_011509256.1 | 3 | 259 | |||
GenBank mRNA | AK222979.1 | - | 8 | 583 | ||
AK223023.1 | - | 8 | 583 | |||
AK308042.1 | - | 8 | 583 | |||
AK313651.1 | - | 8 | 589 | |||
BC005190.1 | - | 8 | 636 | |||
BC067822.1 | - | 8 | 588 | 5266813 | ||
BI764401.1 | - | 8 | 669 | |||
U66669.1 | - | 8 | 613 |