Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKD2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258448.1 | NP_001245377.1 | 10 | 1493 | ||
NM_001258449.1 | NP_001245378.1 | 8 | 1256 | |||
NM_001300921.1 | NP_001287850.1 | 10 | 1559 | |||
NM_014386.3 | NP_055201.2 | 10 | 1559 | |||
XM_011543318.2 | XP_011541620.1 | 9 | 1473 | |||
XM_011543321.1 | XP_011541623.1 | 10 | 1493 | |||
XM_017009343.1 | XP_016864832.1 | 10 | 1575 | |||
XM_017009344.1 | XP_016864833.1 | 9 | 1683 | |||
XM_017009345.1 | XP_016864834.1 | 9 | 1353 | |||
GenBank mRNA | AF118125.1 | - | 10 | 1526 | ||
AF182034.1 | - | 10 | 1529 | |||
AK301924.1 | - | 10 | 1462 | |||
BC044581.1 | - | 8 | 1255 | 4830805 |