Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014391.2 | NP_055206.2 | 7 | 964 | ||
GenBank mRNA | AK290960.1 | - | 7 | 776 | ||
AY903446.1 | - | 7 | 733 | |||
BC018667.1 | - | 7 | 780 | 4245591 | ||
X83703.1 | - | 7 | 965 |