Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164380.1 | NP_001157852.1 | 13 | 1784 | ||
NM_001164381.1 | NP_001157853.1 | 12 | 1653 | |||
NM_001164382.1 | NP_001157854.1 | 11 | 1493 | |||
NM_001164383.1 | NP_001157855.1 | 8 | 1178 | |||
NM_001164384.1 | NP_001157856.1 | 11 | 1717 | |||
NM_001164385.1 | NP_001157857.1 | 10 | 1474 | |||
NM_014393.2 | NP_055208.2 | 12 | 1653 | |||
GenBank mRNA | AB210014.1 | - | 12 | 1549 | ||
AF459097.1 | - | 11 | 1503 | |||
AF459098.1 | - | 10 | 1372 | |||
AK001576.1 | - | 5 | 679 | |||
AK002152.1 | - | 13 | 1697 | |||
AK023314.1 | - | 4 | 556 | |||
AK290141.1 | - | 11 | 1717 | |||
AK293496.1 | - | 8 | 1091 | |||
AK293729.1 | - | 11 | 1543 | |||
AK294466.1 | - | 10 | 1426 | |||
AK303104.1 | - | 11 | 1406 | |||
AK307841.1 | - | 8 | 1114 | |||
AK314617.1 | - | 12 | 1566 | |||
BC008369.1 | - | 12 | 1566 | 3682739 | ||
BC110447.1 | - | 10 | 1390 | 40009098 | ||
CR627442.1 | - | 8 | 1160 | |||
Y19062.1 | - | 12 | 1548 |