Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289036.1 | NP_001275965.1 | 7 | 914 | ||
NM_001318522.1 | NP_001305451.1 | 5 | 630 | |||
NM_014410.4 | NP_055225.1 | 5 | 669 | |||
NM_199167.1 | NP_954636.1 | 5 | 595 | |||
XM_005258103.3 | XP_005258160.1 | 7 | 819 | |||
XM_006722322.2 | XP_006722385.1 | 6 | 787 | |||
XM_011525648.2 | XP_011523950.1 | 7 | 862 | |||
XM_011525649.1 | XP_011523951.1 | 7 | 862 | |||
XM_011525651.2 | XP_011523953.1 | 6 | 1584 | |||
XM_011525652.1 | XP_011523954.1 | 7 | 938 | |||
XM_011525653.2 | XP_011523955.1 | 5 | 559 | |||
XM_017025707.1 | XP_016881196.1 | 8 | 2324 | |||
GenBank mRNA | AF395889.1 | - | 5 | 584 | ||
AK299319.1 | - | 7 | 889 | |||
BC025381.2 | - | 5 | 669 | 4821299 | ||
D63813.1 | - | 5 | 575 |