Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289036.1 | NP_001275965.1 | 10, 11 | 1792 | ||
NM_001318522.1 | NP_001305451.1 | 8, 9 | 1508 | |||
NM_014410.4 | NP_055225.1 | 8, 9 | 1547 | |||
NM_199167.1 | NP_954636.1 | 8, 9 | 1473 | |||
XM_005258103.3 | XP_005258160.1 | 10, 11 | 1697 | |||
XM_006722322.2 | XP_006722385.1 | 9, 10 | 1665 | |||
XM_011525649.1 | XP_011523951.1 | 10, 11 | 1740 | |||
XM_011525651.2 | XP_011523953.1 | 9, 10 | 2462 | |||
XM_011525652.1 | XP_011523954.1 | 10, 11 | 1816 | |||
XM_017025707.1 | XP_016881196.1 | 11, 12 | 3202 | |||
GenBank mRNA | AF395889.1 | - | 8, 9 | 1462 | ||
AK299319.1 | - | 10, 11 | 1767 | |||
BC025381.2 | - | 8, 9 | 1547 | 4821299 | ||
BX108184.1 | - | 1, 2 | 212 | |||
D63813.1 | - | 8, 9 | 1453 |