Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DKK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014420.2 | NP_055235.1 | 2, 3 | 380 | ||
XM_011544488.2 | XP_011542790.1 | 3, 4 | 955 | |||
XM_017013316.1 | XP_016868805.1 | 3, 4 | 678 | |||
GenBank mRNA | AF177397.1 | - | 2, 3 | 386 | ||
AK301609.1 | - | 2, 3 | 450 | |||
BC107046.2 | - | 2, 3 | 307 | 40003485 | ||
BC107047.2 | - | 2, 3 | 307 | 40003487 | ||
BX105775.1 | - | 2, 3 | 380 |