Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DKK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014421.2 | NP_055236.1 | 4 | 2404 | ||
GenBank mRNA | AF177395.1 | - | 4 | 2416 | ||
AY358414.1 | - | 4 | 1909 |