Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INVS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318381.1 | NP_001305310.1 | 5 | 729 | ||
NM_001318382.1 | NP_001305311.1 | 4 | 641 | |||
NM_014425.4 | NP_055240.2 | 4 | 641 | |||
NR_134606.1 | 4 | 641 | ||||
GenBank mRNA | AF039217.2 | - | 4 | 592 | ||
AF084367.1 | - | 3 | 411 | |||
AK304851.1 | - | 5 | 651 | |||
AK312795.1 | - | 4 | 583 | |||
BC041665.1 | - | 4 | 597 | |||
BC056897.1 | - | 6 | 764 | |||
BC111761.1 | - | 3 | 424 | 40012263 | ||
BC112351.1 | - | 3 | 424 | 40012265 |