Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SESN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014454.2 | NP_055269.1 | 1 | 510 | ||
GenBank mRNA | AF033122.1 | - | 1 | 89 | ||
AK290157.1 | - | 1 | 317 | |||
BC112036.1 | - | 1 | 142 | 8327504 | ||
BC113569.1 | - | 1 | 130 | 8322621 |