Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178010.2 | NP_001171481.1 | 17, 18 | 1800 | ||
NM_001178011.2 | NP_001171482.1 | 15, 16 | 1566 | |||
NM_003504.4 | NP_003495.1 | 16, 17 | 1704 | |||
XM_005261285.2 | XP_005261342.1 | 18, 19 | 1893 | |||
XM_011530415.1 | XP_011528717.1 | 17, 18 | 1797 | |||
XM_011530418.2 | XP_011528720.1 | 16, 17 | 1860 | |||
GenBank mRNA | AF053074.1 | - | 16, 17 | 1585 | ||
AF062495.1 | - | 16, 17 | 1577 | |||
AF081535.1 | - | 16, 17 | 1592 | |||
AJ223728.1 | - | 16, 17 | 1591 | |||
AK293123.1 | - | 15, 16 | 1501 | |||
AK293338.1 | - | 17, 18 | 1697 | |||
AY358971.1 | - | 16, 17 | 1627 | |||
BC006232.2 | - | 16, 17 | 1622 | 3946493 | ||
BC010022.2 | - | 16, 17 | 1607 | 2964592 | ||
BT006792.1 | - | 16, 17 | 1561 | |||
CT841513.1 | - | 16, 17 | 1565 |