Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317893.1 | NP_001304822.1 | 3 | 642 | ||
NM_014506.2 | NP_055321.1 | 3 | 642 | |||
NR_133948.1 | 3 | 783 | ||||
NR_134113.1 | 3 | 783 | ||||
XM_005251927.3 | XP_005251984.1 | 3 | 637 | |||
GenBank mRNA | AF007872.1 | - | 2 | 363 | ||
AF317129.1 | - | 3 | 632 | |||
AI568476.1 | - | 2 | 339 | |||
BC015578.2 | - | 3 | 587 | 4589479 | ||
CN262366.1 | - | 2 | 432 |