Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317893.1 | NP_001304822.1 | 4 | 1152 | ||
NM_001317894.1 | NP_001304823.1 | 3 | 976 | |||
NM_014506.2 | NP_055321.1 | 5 | 2165 | |||
NR_133948.1 | 5 | 2306 | ||||
NR_134027.1 | 3 | 1723 | ||||
NR_134113.1 | 5 | 1421 | ||||
XM_005251927.3 | XP_005251984.1 | 5 | 1275 | |||
GenBank mRNA | AF007872.1 | - | 4 | 1887 | ||
AF317129.1 | - | 5 | 2155 | |||
BC006480.1 | - | 1 | 978 | |||
BC015578.2 | - | 5 | 2110 | 4589479 |