Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014548.3 | NP_055363.1 | 7 | 900 | ||
XM_017022089.1 | XP_016877578.1 | 7 | 864 | |||
GenBank mRNA | AF177169.1 | - | 7 | 835 | ||
AK292733.1 | - | 7 | 779 | |||
BC036184.1 | - | 8 | 946 | 4815047 | ||
BC064961.1 | - | 7 | 848 | 6141541 |