Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003551.2 | NP_003542.1 | 6 | 773 | ||
XM_006714716.3 | XP_006714779.1 | 4 | 581 | |||
XM_017009945.1 | XP_016865434.1 | 5 | 1538 | |||
GenBank mRNA | AF067724.1 | - | 6 | 743 | ||
BC026182.2 | - | 6 | 745 | 5270050 | ||
U90450.1 | - | 6 | 719 | |||
Y14992.1 | - | 5 | 709 |