Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083893.1 | NP_001077362.1 | 10 | 1494 | ||
NM_014574.3 | NP_055389.3 | 8 | 1242 | |||
XM_005267569.3 | XP_005267626.1 | 9 | 1510 | |||
XM_005267570.3 | XP_005267627.1 | 9 | 1480 | |||
GenBank mRNA | AF243424.1 | - | 9 | 923 | ||
AK290229.1 | - | 8 | 1220 | |||
AK314041.1 | - | 8 | 1208 | |||
BC126221.1 | - | 8 | 1160 | 8991937 | ||
BC132673.1 | - | 8 | 1160 | 40146695 | ||
BM453981.1 | - | 2 | 136 | |||
U17989.1 | - | 8 | 1230 |