Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278233.1 | NP_001265162.1 | 5 | 1497 | ||
NM_001278234.1 | NP_001265163.1 | 5 | 1330 | |||
NM_014583.3 | NP_055398.1 | 6 | 1586 | |||
XM_011533649.1 | XP_011531951.1 | 7 | 1641 | |||
GenBank mRNA | AF169284.1 | - | 6 | 1557 | ||
AK022176.1 | - | 6 | 1568 | |||
AK074448.1 | - | 1 | 2676 | |||
BC000646.2 | - | 6 | 1556 | 3343074 |