Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERO1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014584.2 | NP_055399.1 | 7 | 984 | ||
XM_011536684.2 | XP_011534986.1 | 7 | 803 | |||
GenBank mRNA | AF081886.1 | - | 7 | 760 | ||
AF123887.1 | - | 5 | 286 | |||
AK292839.1 | - | 7 | 582 | |||
AY358463.1 | - | 7 | 706 | |||
BC008674.1 | - | 7 | 756 | 3866930 | ||
BC012941.1 | - | 7 | 767 | 3868538 |