Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014600.2 | NP_055415.1 | 6 | 3582 | ||
XM_011532806.2 | XP_011531108.1 | 4 | 2546 | |||
GenBank mRNA | AF214736.1 | - | 6 | 3259 |