Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014601.3 | NP_055416.2 | 6 | 3243 | ||
GenBank mRNA | AF454952.1 | - | 5 | 3057 | ||
AK026134.1 | - | 1 | 1561 | |||
AK074701.1 | - | 7 | 2279 | |||
AK097126.1 | - | 6 | 2574 | |||
AK097228.1 | - | 5 | 2567 | |||
BC014445.1 | - | 6 | 3159 | 4908085 |