Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199579.1 | NP_001186508.1 | 7, 8 | 1045 | ||
NM_001199580.1 | NP_001186509.1 | 8, 9 | 981 | |||
NM_015374.2 | NP_056189.1 | 7, 8 | 982 | |||
XM_011530104.2 | XP_011528406.1 | 6, 7 | 839 | |||
XM_011530105.2 | XP_011528407.1 | 7, 8 | 706 | |||
XM_017028748.1 | XP_016884237.1 | 7, 8 | 977 | |||
GenBank mRNA | AB014568.1 | - | 7, 8 | 764 | ||
AK295787.1 | - | 7, 8 | 706 | |||
AK304188.1 | - | 8, 9 | 841 | |||
AY682988.1 | - | 7, 8 | 763 | |||
BC094797.1 | - | 7, 8 | 1002 | 4799090 | ||
BC111549.1 | - | 6, 7 | 711 | 40008751 | ||
BC111717.1 | - | 6, 7 | 711 | 40008756 | ||
BX537962.1 | - | 8, 9 | 973 | |||
CR456474.1 | - | 6, 7 | 709 |