Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100819.2 | NP_001094289.1 | 6 | 436 | ||
NM_001204094.1 | NP_001191023.1 | 7 | 453 | |||
NM_015387.4 | NP_056202.2 | 7 | 499 | |||
NM_199482.3 | NP_955776.1 | 7 | 509 | |||
GenBank mRNA | AB015441.1 | - | 7 | 458 | ||
AF093825.1 | - | 7 | 474 | |||
AF151853.1 | - | 7 | 474 | |||
AF250319.1 | - | 7 | 484 | |||
AJ580638.1 | - | 7 | 455 | |||
AK027043.1 | - | 7 | 496 | |||
AK292938.1 | - | 7 | 495 | |||
AK297294.1 | - | 370 | ||||
AK297514.1 | - | 6 | 395 | |||
AK307497.1 | - | 6 | 367 | |||
AL080070.1 | - | 7 | 468 | |||
BC005237.1 | - | 7 | 499 | 3996585 | ||
CR457371.1 | - | 7 | 455 | |||
DC298610.1 | - | 7 | 509 | |||
DC350915.1 | - | 6 | 432 | |||
DC375649.1 | - | 7 | 455 |
Gene Symbol: HSPE1-MOB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202485.1 | NP_001189414.1 | 8 | 1003 |