Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUMF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130069.2 | NP_001123541.1 | 3 | 418 | ||
NM_001146333.1 | NP_001139805.1 | 2 | 261 | |||
NM_015411.2 | NP_056226.2 | 3 | 418 | |||
XM_011515254.2 | XP_011513556.1 | 3 | 418 | |||
XM_017011939.1 | XP_016867428.1 | 3 | 418 | |||
GenBank mRNA | AK075477.1 | - | 3 | 377 | ||
AK297793.1 | - | 3 | 332 | |||
AK298605.1 | - | 3 | 358 | |||
AK300488.1 | - | 2 | 220 | |||
AY323911.1 | - | 3 | 330 | |||
AY359103.1 | - | 3 | 348 | |||
BC000224.1 | - | 2 | 153 | |||
BC006159.1 | - | 3 | 316 | |||
BC015600.2 | - | 3 | 315 | |||
BC084539.1 | - | 3 | 348 | 6599080 | ||
BC111092.1 | - | 3 | 371 | 40068651 | ||
BM423367.1 | - | 3 | 320 | |||
BP256067.1 | - | 1 | 60 | |||
CF131206.1 | - | 3 | 379 | |||
CR936757.1 | - | 3 | 361 | |||
DA130450.1 | - | 3 | 396 | |||
DA924174.1 | - | 3 | 419 | |||
KU178582.1 | - | 2 | 243 | |||
KU178583.1 | - | 2 | 243 |