Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015417.4 | NP_056232.2 | 7 | 958 | ||
XM_005260683.4 | XP_005260740.1 | 7 | 917 | |||
GenBank mRNA | AL080154.1 | - | 6 | 1084 | ||
BC022476.1 | - | 5 | 1262 | 4816764 | ||
EF560745.1 | - | 7 | 956 |