Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042452.1 | NP_001035917.1 | 10 | 1425 | ||
NM_001042453.1 | NP_001035918.1 | 11 | 1671 | |||
NM_016542.3 | NP_057626.2 | 12 | 1902 | |||
XM_011531349.1 | XP_011529651.1 | 13 | 1875 | |||
XM_011531350.1 | XP_011529652.1 | 12 | 1948 | |||
GenBank mRNA | AB040057.2 | - | 12 | 1821 | ||
AF231012.1 | - | 12 | 1790 | |||
AK000556.1 | - | 8 | 1234 | |||
AK027589.1 | - | 7 | 1119 | |||
AK074837.1 | - | 7 | 1155 | |||
AK075107.1 | - | 11 | 1583 | |||
AK075150.1 | - | 11 | 1745 | |||
BC013856.2 | - | 1 | 387 | |||
BC098315.1 | - | 12 | 1807 | 40026349 | ||
BC099843.1 | - | 12 | 1807 | 40026348 | ||
BC103503.1 | - | 12 | 1807 | 40026350 |