Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNOT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256741.1 | NP_001243670.1 | 7 | 997 | ||
NM_001256742.1 | NP_001243671.1 | 7 | 946 | |||
NM_015442.2 | NP_056257.1 | 7 | 997 | |||
NR_046352.1 | 6 | 835 | ||||
XM_006713084.3 | XP_006713147.1 | 7 | 953 | |||
XM_006713085.3 | XP_006713148.1 | 7 | 953 | |||
XM_011533566.2 | XP_011531868.1 | 7 | 953 | |||
XM_011533567.2 | XP_011531869.1 | 3 | 214 | |||
XM_017006109.1 | XP_016861598.1 | 7 | 997 | |||
XM_017006110.1 | XP_016861599.1 | 7 | 997 | |||
XM_017006111.1 | XP_016861600.1 | 7 | 997 | |||
XM_017006112.1 | XP_016861601.1 | 7 | 997 | |||
GenBank mRNA | AK021695.1 | - | 5 | 431 | ||
AK022576.1 | - | 7 | 799 | |||
AK022952.1 | - | 7 | 936 | |||
AK023227.1 | - | 7 | 954 | |||
AK027026.1 | - | 5 | 557 | |||
AK297085.1 | - | 6 | 572 | |||
AK298693.1 | - | 6 | 635 | |||
AK302196.1 | - | 7 | 938 | |||
AK309670.1 | - | 6 | 760 | |||
BC002928.2 | - | 7 | 899 | 3954507 | ||
BC002931.2 | - | 7 | 905 | 3955575 |