Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNOT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256741.1 | NP_001243670.1 | 11 | 1606 | ||
NM_001256742.1 | NP_001243671.1 | 11 | 1555 | |||
NM_015442.2 | NP_056257.1 | 11 | 1606 | |||
NR_046352.1 | 10 | 1444 | ||||
XM_006713084.3 | XP_006713147.1 | 11 | 1562 | |||
XM_006713085.3 | XP_006713148.1 | 11 | 1562 | |||
XM_011533566.2 | XP_011531868.1 | 11 | 1562 | |||
XM_011533567.2 | XP_011531869.1 | 7 | 823 | |||
XM_017006109.1 | XP_016861598.1 | 11 | 1606 | |||
XM_017006110.1 | XP_016861599.1 | 11 | 1606 | |||
XM_017006111.1 | XP_016861600.1 | 11 | 1606 | |||
XM_017006112.1 | XP_016861601.1 | 11 | 1606 | |||
GenBank mRNA | AK021695.1 | - | 9 | 1040 | ||
AK022576.1 | - | 11 | 1408 | |||
AK022952.1 | - | 11 | 1545 | |||
AK023227.1 | - | 11 | 1563 | |||
AK027026.1 | - | 8 | 1048 | |||
AK297085.1 | - | 10 | 1181 | |||
AK298693.1 | - | 10 | 1244 | |||
AK302196.1 | - | 11 | 1547 | |||
AK309670.1 | - | 10 | 1369 | |||
BC002928.2 | - | 11 | 1508 | 3954507 | ||
BC002931.2 | - | 11 | 1514 | 3955575 |