Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEMIN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252156.1 | NP_001239085.1 | 11 | 1601 | ||
NM_015465.4 | NP_056280.2 | 11 | 1604 | |||
GenBank mRNA | AK022748.1 | - | 11 | 1551 | ||
AK074066.2 | - | 11 | 1576 | |||
AK315101.1 | - | 11 | 1589 | |||
AL117665.1 | - | 6 | 666 | |||
AY063750.1 | - | 11 | 1521 | |||
BC036894.1 | - | 11 | 1584 | |||
BC040931.1 | - | 11 | 1574 | |||
BC113614.1 | - | 11 | 1560 | 8322666 | ||
BC143724.1 | - | 11 | 1557 | 9052238 |