Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304482.1 | NP_001291411.1 | 17 | 1810 | ||
NM_015466.3 | NP_056281.1 | 18 | 1938 | |||
XM_005265031.2 | XP_005265088.2 | 18 | 1889 | |||
GenBank mRNA | AB025194.1 | - | 18 | 1886 | ||
AB040904.2 | - | 18 | 1938 | |||
AF290614.1 | - | 18 | 1884 | |||
AK055157.1 | - | 18 | 1909 | |||
AK289502.1 | - | 18 | 1887 | |||
AK295877.1 | - | 18 | 1858 | |||
AK295888.1 | - | 17 | 1801 | |||
AK299906.1 | - | 17 | 1781 | |||
BC027711.2 | - | 7 | 877 | |||
BC089042.1 | - | 18 | 1856 | 6579163 |