Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPSNAP3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015469.1 | NP_056284.1 | 4 | 613 | ||
GenBank mRNA | AF161417.1 | - | 4 | 594 | ||
AK024015.1 | - | 4 | 574 | |||
AK301410.1 | - | 4 | 604 | |||
AL117557.1 | - | 4 | 557 | |||
BC005935.1 | - | 4 | 613 | 4064889 | ||
BC045830.1 | - | 4 | 553 |