Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282433.1 | NP_001269362.1 | 6 | 1226 | ||
NM_015515.4 | NP_056330.3 | 7 | 1754 | |||
XM_005257200.4 | XP_005257257.1 | 6 | 807 | |||
XM_011524595.1 | XP_011522897.1 | 6 | 797 | |||
GenBank mRNA | AF102848.1 | - | 6 | 1182 | ||
AK002047.1 | - | 6 | 1166 | |||
AK225866.1 | - | 5, 6 | 1230 | |||
AK289449.1 | - | 6 | 1249 | |||
AK302109.1 | - | 6 | 1035 | |||
AK310880.1 | - | 6 | 1338 | |||
AK314604.1 | - | 6 | 966 | |||
AL117538.1 | - | 5 | 1010 | |||
BC028356.2 | - | 7 | 1534 | 4838347 | ||
CV373638.1 | - | 3 | 442 |