Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GORASP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201428.1 | NP_001188357.1 | 5 | 520 | ||
NM_015530.4 | NP_056345.3 | 5 | 1275 | |||
XM_006712408.2 | XP_006712471.1 | 5 | 598 | |||
GenBank mRNA | AK023082.1 | - | 5 | 547 | ||
AK023201.1 | - | 5 | 560 | |||
AK027349.1 | - | 5 | 474 | |||
AK125761.1 | - | 5 | 1275 | |||
AK225810.1 | - | 5 | 557 | |||
AK293640.1 | - | 3, 4 | 407 | |||
AK296698.1 | - | 6 | 584 | |||
AK298020.1 | - | 5 | 554 | |||
AK298771.1 | - | 3 | 141 | |||
AL117430.1 | - | 5 | 486 | |||
BC007770.2 | - | 5 | 509 | 4298251 |