Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSMF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130969.1 | NP_001124441.1 | 4 | 925 | ||
NM_001130970.1 | NP_001124442.1 | 4 | 925 | |||
NM_001130971.1 | NP_001124443.1 | 4 | 925 | |||
NM_001178064.1 | NP_001171535.1 | 4 | 925 | |||
NM_015537.4 | NP_056352.3 | 4 | 925 | |||
XM_005266061.4 | XP_005266118.1 | 4 | 948 | |||
XM_005266062.4 | XP_005266119.1 | 4 | 948 | |||
XM_011518496.2 | XP_011516798.1 | 4 | 948 | |||
GenBank mRNA | AK027474.1 | - | 2 | 196 | ||
AK074602.1 | - | 4 | 893 | |||
AK292030.1 | - | 1 | 66 | |||
AK294320.1 | - | 2 | 160 | |||
AK303703.1 | - | 2 | 116 | |||
AK316046.1 | - | 2 | 310 | |||
AL117660.2 | - | 1 | 19 | |||
AY255128.1 | - | 4 | 925 | |||
AY255129.1 | - | 4 | 925 | |||
AY255130.1 | - | 4 | 925 | |||
AY255131.1 | - | 4 | 925 | |||
AY255132.1 | - | 4 | 925 | |||
BC072412.1 | - | 4 | 910 | 6165009 | ||
BC110498.2 | - | 4 | 792 | 40007676 |